Gene Transcripts
- Gene Symbol
- PSMD13
- Entrez Gene ID
- 5719
- Gene Name
- proteasome 26S subunit, non-ATPase 13
- Gene Aliases
- HSPC027, Rpn9, S11, p40.5
- Chromosome Location
- Chr. 11: 236675 - 252984 on Build GRCh38
- UniGene ID
- Hs.134688
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK092845.1 |
- |
1 |
1464 |
|
View Full Product Details