Gene Transcripts
- Gene Symbol
- MTMR9
- Entrez Gene ID
- 66036
- Gene Name
- myotubularin related protein 9
- Gene Aliases
- C8orf9, LIP-STYX, MTMR8
- Chromosome Location
- Chr. 8: 11284491 - 11328146 on Build GRCh38
- UniGene ID
- Hs.591395
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK022071.1 |
- |
1 |
262 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BQ441275.1 |
- |
2 |
432 |
|
Chr. 8 - 11295198 - 11300609 [+] |
GenBankmRNA |
DA829171.1 |
- |
2 |
471 |
|
Chr. 8 - 11295157 - 11300667 [+] |
GenBankmRNA |
AU120884.1 |
- |
1 |
262 |
|
Chr. 8 - 11300086 - 11300812 [+] |
View Full Product Details