Gene Transcripts
- Gene Symbol
- Mmp28
- Entrez Gene ID
- 118453
- Gene Name
- matrix metallopeptidase 28 (epilysin)
- Gene Aliases
- D130023P12
- Chromosome Location
- Chr. 11: 83441241 - 83463064 on Build GRCm38
- UniGene ID
- Mm.264648
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK087705.1 |
- |
1 |
961 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB553843.2 |
- |
1 |
151 |
|
Chr. 11 - 83461538 - 83462125 [-] |
View Full Product Details