Gene Transcripts
- Gene Symbol
- Azin1
- Entrez Gene ID
- 54375
- Gene Name
- antizyme inhibitor 1
- Gene Aliases
- 1700085L02Rik, AZI, Oazi, Oazin
- Chromosome Location
- Chr. 15: 38487430 - 38519287 on Build GRCm38
- UniGene ID
- Mm.250214
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK007010.1 |
- |
|
1065 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BX516018.1 |
- |
1 |
416 |
|
Chr. 15 - 38519438 - 38519940 [+] |
View Full Product Details