Gene Transcripts
- Gene Symbol
- Acsl5
- Entrez Gene ID
- 433256
- Gene Name
- acyl-CoA synthetase long-chain family member 5
- Gene Aliases
- 1700030F05Rik, ACS2, ACS5, Facl5
- Chromosome Location
- Chr. 19: 55251862 - 55296628 on Build GRCm38
- UniGene ID
- Mm.292056
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK054476.1 |
- |
|
1399 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BG923724.1 |
- |
1 |
356 |
|
Chr. 19 - 55295959 - 55296548 [-] |
GenBankmRNA |
BB251700.1 |
- |
1 |
306 |
|
Chr. 19 - 55296190 - 55296521 [-] |
GenBankmRNA |
BB557586.2 |
- |
1 |
639 |
|
Chr. 19 - 55296194 - 55296853 [-] |
GenBankmRNA |
AK163931.1 |
- |
|
1394 |
|
Chr. - - [-] |
View Full Product Details