Gene Transcripts
- Gene Symbol
- Lrrc52
- Entrez Gene ID
- 240899
- Gene Name
- leucine rich repeat containing 52
- Gene Aliases
- 4930413P14Rik
- Chromosome Location
- Chr. 1: 167445675 - 167466780 on Build GRCm38
- UniGene ID
- Mm.159799
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015134.1 |
- |
|
1880 |
|
View Full Product Details