Gene Transcripts

Gene Symbol
ANKS6
Entrez Gene ID
203286
Gene Name
ankyrin repeat and sterile alpha motif domain containing 6
Gene Aliases
ANKRD14, NPHP16, PKDR1, SAMD6
Chromosome Location
Chr. 9: 98732009 - 98796542 on Build GRCh38
UniGene ID
Hs.406890
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173551.4 NP_775822.3 6 1312
XM_005251793.4 XP_005251850.1 6 1299
XM_005251794.4 XP_005251851.1 6 1299
XM_006716998.3 XP_006717061.1 6 1299
XM_006716999.2 XP_006717062.1 6 2384
XM_011518357.2 XP_011516659.1 6 3381
XM_017014445.1 XP_016869934.1 6 1299
XM_017014446.1 XP_016869935.1 5 3260
GenBank mRNA AK126614.1 - 4 366
AK127702.1 - 5 899
BC064367.1 - 3 299 5562895
CR749467.1 - 6 983
CR749472.1 - 4 663

View Full Product Details