Gene Transcripts
- Gene Symbol
- LINC01559
- Entrez Gene ID
- 283422
- Gene Name
- long intergenic non-protein coding RNA 1559
- Gene Aliases
- C12orf36
- Chromosome Location
- Chr. 12: 13370671 - 13376745 on Build GRCh38
- UniGene ID
- Hs.448717
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_036555.1 |
|
3 |
884 |
|
GenBank mRNA |
AK091129.1 |
- |
3 |
850 |
|
View Full Product Details