Gene Transcripts
- Gene Symbol
- SRP14-AS1
- Entrez Gene ID
- 100131089
- Gene Name
- SRP14 antisense RNA1 (head to head)
- Gene Aliases
- Chromosome Location
- Chr. 15: 40039311 - 40067509 on Build GRCh38
- UniGene ID
- Hs.386000
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000560341.2 |
- |
1 |
80 |
|
Chr. 15 - 40039349 - 40056983 [+] |
GENCODE |
ENST00000504245.5 |
- |
1 |
118 |
|
Chr. 15 - 40039311 - 40067290 [+] |
View Full Product Details