Gene Transcripts
- Gene Symbol
- EYA1
- Entrez Gene ID
- 2138
- Gene Name
- EYA transcriptional coactivator and phosphatase 1
- Gene Aliases
- BOP, BOR, BOS1, OFC1
- Chromosome Location
- Chr. 8: 71197433 - 71548130 on Build GRCh38
- UniGene ID
- Hs.444971
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC035138.1 |
- |
4 |
1596 |
|
BC048982.1 |
- |
4 |
1590 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000523987.4 |
- |
4 |
1589 |
|
Chr. 8 - 71403440 - 71547657 [-] |
View Full Product Details