Gene Transcripts
- Gene Symbol
- TRG-AS1
- Entrez Gene ID
- 100506776
- Gene Name
- T cell receptor gamma locus antisense RNA 1
- Gene Aliases
- TCRGV
- Chromosome Location
- Chr. 7: 38341577 - 38378637 on Build GRCh38
- UniGene ID
- Hs.173034
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040085.1 |
|
1 |
86 |
|
GenBank mRNA |
AK125646.1 |
- |
1 |
48 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000629357.1 |
- |
|
48 |
|
Chr. - - [-] |
GENCODE |
ENST00000629576.1 |
- |
|
86 |
|
Chr. - - [-] |
GenBankmRNA |
DA992081.1 |
- |
1 |
48 |
|
Chr. 7 - 38341615 - 38375712 [+] |
View Full Product Details