Gene Transcripts
- Gene Symbol
- Alpk3
- Entrez Gene ID
- 116904
- Gene Name
- alpha-kinase 3
- Gene Aliases
- AW319487, D330016D04, MAK, Midori, mKIAA1330
- Chromosome Location
- Chr. 7: 81057600 - 81105612 on Build GRCm38
- UniGene ID
- Mm.475768
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_054085.2 |
NP_473426.2
|
1, 2 |
174 |
|
GenBank mRNA |
AF338872.1 |
- |
1, 2 |
174 |
|
View Full Product Details