Gene Transcripts

Gene Symbol
Bud13
Entrez Gene ID
215051
Gene Name
BUD13 homolog (yeast)
Gene Aliases
AV305342, D030060M11Rik
Chromosome Location
Chr. 9: 46283012 - 46298783 on Build GRCm38
UniGene ID
Mm.32648
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146000.2 NP_666112.1 5 1135
GenBank mRNA AK037134.1 - 5 1132
AK038126.1 - 5 1140
AK051047.1 - 5 1140
AK077744.1 - 5 1140
BC025490.1 - 5 1136 5252666

View Full Product Details