Gene Transcripts

Gene Symbol
Baz1a
Entrez Gene ID
217578
Gene Name
bromodomain adjacent to zinc finger domain 1A
Gene Aliases
Acf1, B930060C03, BC065123, Gtl5, Wcrf180, cbp146
Chromosome Location
Chr. 12: 54892600 - 55014371 on Build GRCm38
UniGene ID
Mm.479583
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013815.2 NP_038843.2 26 4624
GenBank mRNA AK047427.1 - 11 2432
AK132694.1 - 9 1820
BC065123.1 - 12 2440

View Full Product Details