Gene Transcripts

Gene Symbol
Acat3
Entrez Gene ID
224530
Gene Name
acetyl-Coenzyme A acetyltransferase 3
Gene Aliases
ACTL
Chromosome Location
Chr. 17: 12923959 - 12940396 on Build GRCm38
UniGene ID
Mm.490312
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153151.3 NP_694791.2 9 1751
GenBank mRNA AK143884.1 - 9 1711
AK149441.1 - 9 1751

View Full Product Details