Gene Transcripts

Gene Symbol
Wdr36
Entrez Gene ID
225348
Gene Name
WD repeat domain 36
Gene Aliases
5730444A13Rik, Ta-wdrp
Chromosome Location
Chr. 18: 32837225 - 32866420 on Build GRCm38
UniGene ID
Mm.38816
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110015.1 NP_001103485.1 9 1117
NM_001110016.1 NP_001103486.1 9 1117
NM_144863.4 NP_659112.2 9 1117
GenBank mRNA AK041763.1 - 9 1037
AK045396.1 - 9 1090
AK077545.1 - 9 1046
AK140019.1 - 9 1042
AK155965.1 - 9 1016
AK156822.1 - 1118
AK171683.1 - 9 1055
AK172054.1 - 9 1042
AK172377.1 - 9 1047
AK214340.1 - 1 19
BC022132.1 - 9 1089
BC025809.1 - 9 1031

View Full Product Details