Gene Transcripts
- Gene Symbol
- Wdr36
- Entrez Gene ID
- 225348
- Gene Name
- WD repeat domain 36
- Gene Aliases
- 5730444A13Rik, Ta-wdrp
- Chromosome Location
- Chr. 18: 32837225 - 32866420 on Build GRCm38
- UniGene ID
- Mm.38816
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001110015.1 |
NP_001103485.1
|
5, 6 |
707 |
|
NM_001110016.1 |
NP_001103486.1
|
5, 6 |
707 |
|
NM_144863.4 |
NP_659112.2
|
5, 6 |
707 |
|
GenBank mRNA |
AK041763.1 |
- |
5, 6 |
627 |
|
AK045396.1 |
- |
5, 6 |
680 |
|
AK077545.1 |
- |
5, 6 |
637 |
|
AK140019.1 |
- |
5, 6 |
632 |
|
AK155965.1 |
- |
5, 6 |
606 |
|
AK156822.1 |
- |
|
708 |
|
AK171683.1 |
- |
5, 6 |
645 |
|
AK172054.1 |
- |
5, 6 |
632 |
|
AK172377.1 |
- |
5, 6 |
637 |
|
BC022132.1 |
- |
5, 6 |
679 |
|
BC025809.1 |
- |
5, 6 |
621 |
|
BC094453.1 |
- |
5, 6 |
658 |
5068941 |
View Full Product Details