Gene Transcripts

Gene Symbol
Bahd1
Entrez Gene ID
228536
Gene Name
bromo adjacent homology domain containing 1
Gene Aliases
AL022997, AW541238, Gm117, mKIAA0945
Chromosome Location
Chr. 2: 118900332 - 118924528 on Build GRCm38
UniGene ID
Mm.330487
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001045523.1 NP_001038988.1 2 324
GenBank mRNA AK129243.1 - 2 482
AK152597.1 - 2 262
BC100358.1 - 2 486 30848279
BC141046.1 - 1 187 9055658
BC145233.1 - 1 187 9053785
BY261229.1 - 2 324

View Full Product Details