Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153583.10 | NP_705811.8 | 3 | 548 | |
GenBank mRNA | AK036282.1 | - | 3 | 535 | |
AK036571.1 | - | 3 | 537 | ||
AK081002.1 | - | 3 | 535 | ||
AK081605.1 | - | 3 | 526 | ||
AK087258.1 | - | 3 | 538 | ||
AK154130.1 | - | 3 | 541 | ||
BC030861.1 | - | 3 | 525 | 4208700 | |
BC069851.1 | - | 3 | 500 | 6490007 |