Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_146230.3 | NP_666342.1 | 12 | 1601 | |
GenBank mRNA | AK145953.1 | - | 12 | 1530 | |
AK160568.1 | - | 12 | 1448 | ||
AY273812.1 | - | 12 | 1599 | ||
BC019882.1 | - | 12 | 1514 | 5123887 |