Gene Transcripts

Gene Symbol
Pcyt1b
Entrez Gene ID
236899
Gene Name
phosphate cytidylyltransferase 1, choline, beta isoform
Gene Aliases
AW045697, CTTbeta
Chromosome Location
Chr. X: 93654863 - 93749951 on Build GRCm38
UniGene ID
Mm.166467
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177546.2 NP_808214.1 7 1028
NM_211138.1 NP_997593.1 7 1039
GenBank mRNA AK032027.1 - 7 741
AK053577.1 - 7 1013
AK149304.1 - 7 1029
AY189949.1 - 7 876
AY189950.1 - 7 1028
BC048917.1 - 7 910 6309383

View Full Product Details