Gene Transcripts

Gene Symbol
MAPK11
Entrez Gene ID
5600
Gene Name
mitogen-activated protein kinase 11
Gene Aliases
P38B, P38BETA2, PRKM11, SAPK2, SAPK2B, p38-2, p38Beta
Chromosome Location
Chr. 22: 50263713 - 50270393 on Build GRCh38
UniGene ID
Hs.57732
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002751.6 NP_002742.3 11, 12 1123
NR_110887.1 11, 12 1123
XM_011530706.1 XP_011529008.1 9, 10 847
GenBank mRNA AF001008.1 - 11, 12 1022
AF001174.1 - 11, 12 1028
AF031135.1 - 11, 12 1037
AK291845.1 - 11, 12 1074
AK308969.1 - 10, 11 960
BC027933.1 - 11, 12 1080 5199628
CR456514.1 - 11, 12 1024
U53442.1 - 11, 12 1065
U92268.1 - 11, 12 1026
Y14440.1 - 11, 12 1028

View Full Product Details