Gene Transcripts
- Gene Symbol
- Rbfox1
- Entrez Gene ID
- 268859
- Gene Name
- RNA binding protein, fox-1 homolog (C. elegans) 1
- Gene Aliases
- A2bp, A2bp1, Hrnbp1, fox-1
- Chromosome Location
- Chr. 16: 5884793 - 7412480 on Build GRCm38
- UniGene ID
- Mm.370334
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021477.5 |
NP_067452.2
|
9, 10 |
1892 |
|
NM_183188.2 |
NP_899011.2
|
5, 6 |
787 |
|
GenBank mRNA |
AB041596.2 |
- |
8, 9 |
1199 |
|
AF107204.1 |
- |
5, 6 |
575 |
|
AF191501.1 |
- |
5, 6 |
788 |
|
AK134833.1 |
- |
7, 8 |
823 |
|
AY659954.1 |
- |
5, 6 |
563 |
|
AY659955.1 |
- |
5, 6 |
563 |
|
AY659956.1 |
- |
5, 6 |
623 |
|
AY659957.1 |
- |
5, 6 |
623 |
|
BC053009.1 |
- |
9, 10 |
1892 |
|
BC059002.1 |
- |
8, 9 |
1556 |
6821627 |
EF410136.1 |
- |
7, 8 |
943 |
|
View Full Product Details