Gene Transcripts

Gene Symbol
Catsperg1
Entrez Gene ID
320225
Gene Name
catsper channel auxiliary subunit gamma 1
Gene Aliases
A230107C01Rik, Catsperg
Chromosome Location
Chr. 7: 29181532 - 29215631 on Build GRCm38
UniGene ID
Mm.349382
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164658.1 NP_001158130.1 22, 23 2720
GenBank mRNA AK133307.1 - 22, 23 2501
BC172039.1 - 16, 17 1760

View Full Product Details