Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002855.4 | NP_002846.3 | 4 | 983 | |
NM_203285.1 | NP_976030.1 | 4 | 983 | ||
NM_203286.1 | NP_976031.1 | 4 | 983 | ||
GenBank mRNA | AF060231.1 | - | 4 | 967 | |
AF110314.1 | - | 4 | 811 | ||
AY029539.1 | - | 4 | 811 | ||
BC104948.1 | - | 4 | 889 | 8143951 | |
BC113471.1 | - | 4 | 885 | 8322523 | |
BQ188335.1 | - | 2 | 217 | ||
X76400.2 | - | 4 | 802 |