Gene Transcripts

Gene Symbol
Acsbg2
Entrez Gene ID
328845
Gene Name
acyl-CoA synthetase bubblegum family member 2
Gene Aliases
Bgr
Chromosome Location
Chr. 17: 56843103 - 56874504 on Build GRCm38
UniGene ID
Mm.179421
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039114.1 NP_001034203.1 13 2157
GenBank mRNA DQ250679.1 - 13 2157

View Full Product Details