Gene Transcripts

Gene Symbol
Baz2b
Entrez Gene ID
407823
Gene Name
bromodomain adjacent to zinc finger domain, 2B
Gene Aliases
5830435C13Rik, BC053917, D2Ertd794e
Chromosome Location
Chr. 2: 59899363 - 60209846 on Build GRCm38
UniGene ID
Mm.228071
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001182.3 NP_001001182.2 9 1712
GenBank mRNA AK029231.1 - 3 760
AK030867.1 - 8 1628
AK134250.1 - 10 1789
AK147737.1 - 10 1904
AK173180.1 - 10 1810
BC027336.1 - 6 1337
BC150814.1 - 10 1694 9087725

View Full Product Details