Gene Transcripts

Gene Symbol
Plcxd2
Entrez Gene ID
433022
Gene Name
phosphatidylinositol-specific phospholipase C, X domain containing 2
Gene Aliases
AW494639, EG433022
Chromosome Location
Chr. 16: 45959261 - 46010413 on Build GRCm38
UniGene ID
Mm.380993
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134480.1 NP_001127952.1 2 1163
GenBank mRNA BC151059.1 - 2 765
BC151063.1 - 2 765 9087976

View Full Product Details