Gene Transcripts

Gene Symbol
Akirin2
Entrez Gene ID
433693
Gene Name
akirin 2
Gene Aliases
2700059D21Rik, AA114675, AA522011, AU019887, Akirin-2
Chromosome Location
Chr. 4: 34550615 - 34566911 on Build GRCm38
UniGene ID
Mm.389532
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007589.3 NP_001007590.2 3, 4 950
GenBank mRNA BC082305.1 - 3, 4 3310

View Full Product Details