Gene Transcripts

Gene Symbol
Adam39
Entrez Gene ID
546055
Gene Name
a disintegrin and metallopeptidase domain 39
Gene Aliases
1700056P18Rik
Chromosome Location
Chr. 8: 40823009 - 40826861 on Build GRCm38
UniGene ID
Mm.334503
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025380.3 NP_001020551.3 2 1004

View Full Product Details