Gene Transcripts
- Gene Symbol
- Uap1
- Entrez Gene ID
- 107652
- Gene Name
- UDP-N-acetylglucosamine pyrophosphorylase 1
- Gene Aliases
- AA420407, AA437972, AGX-1, AGX-2, AGX1, AgX, ESTM38, SPAG2
- Chromosome Location
- Chr. 1: 170142002 - 170174971 on Build GRCm38
- UniGene ID
- Mm.27969
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AA646755.1 |
- |
1 |
167 |
|
View Full Product Details