Gene Transcripts

Gene Symbol
SALL2
Entrez Gene ID
6297
Gene Name
spalt like transcription factor 2
Gene Aliases
COLB, HSAL2, Sal-2, ZNF795, p150(Sal2)
Chromosome Location
Chr. 14: 21521080 - 21537216 on Build GRCh38
UniGene ID
Hs.416358
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291446.1 NP_001278375.1 3 1531
NM_001291447.1 NP_001278376.1 3 1511
NM_005407.2 NP_005398.2 2 1936
XM_011537064.1 XP_011535366.1 2 1862
XM_011537065.1 XP_011535367.1 2 2008
GenBank mRNA AB002358.2 - 2 1923
AK293574.1 - 3 1217
AK296414.1 - 3 1330
AK299931.1 - 2 1810
BC136528.1 - 2 1671 9020518
BC136529.1 - 2 1671 9020519

View Full Product Details