Gene Transcripts

Gene Symbol
Tomt
Entrez Gene ID
791260
Gene Name
transmembrane O-methyltransferase
Gene Aliases
Comt2, F930017I19Rik
Chromosome Location
Chr. 7: 101899803 - 101906359 on Build GRCm38
UniGene ID
Mm.475469
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081679.1 NP_001075148.1 4 635
NM_001282088.1 NP_001269017.1 4 644
GenBank mRNA AK158024.1 - 645
DQ854743.1 - 4 635
EU627091.1 - 4 644

View Full Product Details