Gene Transcripts

Gene Symbol
Cad
Entrez Gene ID
24240
Gene Name
carbamoyl-phosphate synthetase 2, aspartate transcarbamylase, and dihydroorotase
Gene Aliases
Chromosome Location
Chr. 6: 26657507 - 26680459 on Build Rnor_6.0
UniGene ID
Rn.19416
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105710.2 NP_001099180.2 43 6734

View Full Product Details