Gene Transcripts
- Gene Symbol
- Prp15
- Entrez Gene ID
- 24687
- Gene Name
- proline-rich protein 15
- Gene Aliases
- Prp, Prpb, RATRP13, Rp13
- Chromosome Location
- Chr. 4: 166222418 - 166293492 on Build Rnor_6.0
- UniGene ID
- Rn.9841
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012632.1 |
NP_036764.1
|
4 |
746 |
|
View Full Product Details