Gene Transcripts

Gene Symbol
Akr1c14
Entrez Gene ID
191574
Gene Name
aldo-keto reductase family 1, member C14
Gene Aliases
Akr1c9
Chromosome Location
Chr. 17: 69806095 - 69827110 on Build Rnor_6.0
UniGene ID
Rn.10021
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138547.2 NP_612556.1 8 961
GenBank mRNA BC091123.1 - 8 961
D17310.1 - 8 1149
M64393.1 - 8 960
S57790.1 - 8 919

View Full Product Details