Gene Transcripts

Gene Symbol
Caly
Entrez Gene ID
192349
Gene Name
calcyon neuron-specific vesicular protein
Gene Aliases
Calcyon, Drd1ip
Chromosome Location
Chr. 1: 212537851 - 212549040 on Build Rnor_6.0
UniGene ID
Rn.27756
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190399.1 NP_001177328.1 4 373
NM_138915.1 NP_620270.1 4 432
GenBank mRNA AF303658.1 - 4 432
FQ213422.1 - 4 347

View Full Product Details