Gene Transcripts

Gene Symbol
Bzrap1
Entrez Gene ID
287609
Gene Name
benzodiazepine receptor (peripheral) associated protein 1
Gene Aliases
Chromosome Location
Chr. 10: 75054070 - 75078571 on Build Rnor_6.0
UniGene ID
Rn.183937
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008768165.1 XP_008766387.1 18 3945
XM_213427.7 XP_213427.5 18 3945
GenBank mRNA AF199337.1 - 18 2839
AF199338.1 - 18 2839

View Full Product Details