Gene Transcripts

Gene Symbol
Ormdl2
Entrez Gene ID
288783
Gene Name
ORMDL sphingolipid biosynthesis regulator 2
Gene Aliases
Chromosome Location
Chr. 7: 3242991 - 3246097 on Build Rnor_6.0
UniGene ID
Rn.106721
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105940.1 NP_001099410.1 3 393
XM_006240753.2 XP_006240815.1 3 373
XM_006240754.2 XP_006240816.1 3 293

View Full Product Details