Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005999.2 | NP_005990.1 | 6 | 708 | |
GenBank mRNA | AK313068.1 | - | 6 | 646 | |
BC010376.1 | - | 6 | 666 | 4283594 | |
BC011797.1 | - | 6 | 683 | 4138517 | |
BX537610.1 | - | 5 | 3676 | ||
X95073.1 | - | 6 | 710 |