Gene Transcripts

Gene Symbol
Tbc1d19
Entrez Gene ID
289657
Gene Name
TBC1 domain family, member 19
Gene Aliases
RGD1304958
Chromosome Location
Chr. 14: 59403930 - 59516854 on Build Rnor_6.0
UniGene ID
Rn.229690
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106008.2 NP_001099478.1 10, 11 774
GenBank mRNA BC166575.1 - 10, 11 774

View Full Product Details