Gene Transcripts

Gene Symbol
Acy3
Entrez Gene ID
293653
Gene Name
aspartoacylase (aminocyclase) 3
Gene Aliases
RGD1305394
Chromosome Location
Chr. 1: 219233712 - 219237103 on Build Rnor_6.0
UniGene ID
Rn.22552
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009603.1 NP_001009603.1 2 387
XM_006230733.2 XP_006230795.1 2 396
XM_006230734.2 XP_006230796.1 2 449
XM_008760098.1 XP_008758320.1 2 557
XM_008760099.1 XP_008758321.1 2 449
GenBank mRNA BC088190.1 - 2 387

View Full Product Details