Gene Transcripts

Gene Symbol
AQP8
Entrez Gene ID
343
Gene Name
aquaporin 8
Gene Aliases
AQP-8
Chromosome Location
Chr. 16: 25216917 - 25228932 on Build GRCh38
UniGene ID
Hs.176658
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001169.2 NP_001160.2 2 248
XM_011545822.2 XP_011544124.1 2 291
XM_011545823.2 XP_011544125.1 2 298
GenBank mRNA AB013456.1 - 2 250
AF067797.1 - 2 251
AK222964.1 - 2 213
AK222975.1 - 2 245
BC040630.1 - 2 216 5759105

View Full Product Details