Gene Transcripts

Gene Symbol
Pltp
Entrez Gene ID
296371
Gene Name
phospholipid transfer protein
Gene Aliases
Bpife
Chromosome Location
Chr. 3: 161304469 - 161322600 on Build Rnor_6.0
UniGene ID
Rn.117434
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168543.1 NP_001162015.1 5, 6 745
XM_006235535.2 XP_006235597.1 6, 7 803
XM_008762476.1 XP_008760698.1 6, 7 797

View Full Product Details