Gene Transcripts

Gene Symbol
Pof1b
Entrez Gene ID
302328
Gene Name
premature ovarian failure 1B
Gene Aliases
RGD1560798
Chromosome Location
Chr. X: 84099618 - 84167717 on Build Rnor_6.0
UniGene ID
Rn.141102
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106934.1 NP_001100404.1 12 1379

View Full Product Details