Gene Transcripts

Gene Symbol
Mmgt2
Entrez Gene ID
303211
Gene Name
membrane magnesium transporter 2
Gene Aliases
RGD1311260
Chromosome Location
Chr. 10: 48986163 - 48989596 on Build Rnor_6.0
UniGene ID
Rn.23970
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013967.1 NP_001013989.1 2 622
GenBank mRNA BC061881.1 - 2 622

View Full Product Details