Gene Transcripts

Gene Symbol
Armc6
Entrez Gene ID
306352
Gene Name
armadillo repeat containing 6
Gene Aliases
RGD1306691
Chromosome Location
Chr. 16: 20939311 - 20949732 on Build Rnor_6.0
UniGene ID
Rn.218279
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107306.1 NP_001100776.1 2 478
GenBank mRNA BC166821.1 - 134

View Full Product Details