Gene Transcripts

Gene Symbol
Akr1e2
Entrez Gene ID
307091
Gene Name
aldo-keto reductase family 1, member E2
Gene Aliases
Akr1cl2, Akr1e1
Chromosome Location
Chr. 17: 69365440 - 69379944 on Build Rnor_6.0
UniGene ID
Rn.108718
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008342.1 NP_001008343.1 6 613
XM_008771865.1 XP_008770087.1 5 516
GenBank mRNA BC086397.1 - 6 613
FQ212583.1 - 6 626

View Full Product Details