Gene Transcripts

Gene Symbol
Rhobtb1
Entrez Gene ID
309722
Gene Name
Rho-related BTB domain containing 1
Gene Aliases
Chromosome Location
Chr. 20: 20638579 - 20714837 on Build Rnor_6.0
UniGene ID
Rn.3782
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107622.1 NP_001101092.1 6 1242
XM_008772882.1 XP_008771104.1 6 3092

View Full Product Details