Gene Transcripts

Gene Symbol
Rictor
Entrez Gene ID
310131
Gene Name
RPTOR independent companion of MTOR, complex 2
Gene Aliases
Fyb
Chromosome Location
Chr. 2: 56013898 - 56105818 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008760806.1 XP_008759028.1 5 1761
XM_008760807.1 XP_008759029.1 8 671
XM_008760808.1 XP_008759030.1 7 634

View Full Product Details